Order Entry
Switzerland
ContactUsLinkComponent
Herbicide reference standard, Linuron (N-(3,4-Dichlorophenyl)-N'-methoxy-N'-methylurea) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Herbicide reference standard, Linuron (N-(3,4-Dichlorophenyl)-N'-methoxy-N'-methylurea) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL14640000AL
Supplier:  EHRENSTORFER
CAS Number:  
Herbicide reference standard, Linuron (N-(3,4-Dichlorophenyl)-N'-methoxy-N'-methylurea) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL14640000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    330-55-2
  • Description:
    Linuron (N-(3,4-Dichlorophenyl)-N'-methoxy-N'-methylurea) 10 µg/ml in acetonitrile
  • Formula:
    C₉H₁₀Cl₂N₂O₂
  • Description:
    Linuron (N-(3,4-Dichlorophenyl)-N'-methoxy-N'-methylurea)

Specifications

About this item