Order Entry
Switzerland
ContactUsLinkComponent
Pesticide reference standard, Acephate 100 µg/ml in acetonitrile, CIL
Pesticide reference standard, Acephate 100 µg/ml in acetonitrile, CIL
Catalog # PROOCILULM72631.2
CAS Number:  
Pesticide reference standard, Acephate 100 µg/ml in acetonitrile, CIL
Catalog # PROOCILULM72631.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    30560-19-1
  • Description:
    Acephate 100 µg/ml in acetonitrile
  • Formula:
    C₄H₁₀NO₃PS
  • Description:
    Acephate

Specifications

About this item