Order Entry
Switzerland
ContactUsLinkComponent
 
Pesticide reference standard, 1,8-Dichloronaphthalene 100 µg/ml in n-nonane, CIL
  PROOCIL-ECN-2623
 :  
Pesticide reference standard, 1,8-Dichloronaphthalene 100 µg/ml in n-nonane, CIL
  PROOCIL-ECN-2623
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    2050-74-0
  • Description:
    1,8-Dichloronaphthalene 100 µg/ml in n-nonane
  • Environmentally Preferable:
  • Formula:
    C₁₀H₆Cl₂
  • Description:
    1,8-Dichloronaphthalene