SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standards
Organic Standards
Pesticide reference standard, β-BHC 50 µg/ml in n-nonane, CIL
Pesticide reference standard, β-BHC 50 µg/ml in n-nonane, CIL
PROOCILULM61321.2
:
LGC Standards PROMOCHEM
:
Pesticide reference standard, β-BHC 50 µg/ml in n-nonane, CIL
PROOCILULM61321.2
:
LGC Standards PROMOCHEM
:
Pk:
1
CAS No.:
319-85-7
Description:
β-BHC 50 µg/ml in n-nonane
Formula:
C₆H₆Cl₆
Description:
β-BHC