SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standards
Organic Standards
Pesticide reference standard, ε-BHC 10 µg/ml in methanol
Pesticide reference standard, ε-BHC 10 µg/ml in methanol
PROONE7215M
:
LGC Standards PROMOCHEM
:
Pesticide reference standard, ε-BHC 10 µg/ml in methanol
PROONE7215M
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
1,5 mL
CAS No.:
6108-10-7
Description:
ε-BHC 10 µg/ml in methanol
Formula:
C₆H₆Cl₆
Description:
ε-BHC