Order Entry
Switzerland
ContactUsLinkComponent
Pesticide reference standard, 3,4,5-Trichlorophenol 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Pesticide reference standard, 3,4,5-Trichlorophenol 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA17774800AL
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 3,4,5-Trichlorophenol 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA17774800AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    609-19-8
  • Description:
    3,4,5-Trichlorophenol 100 µg/ml in acetonitrile
  • Formula:
    C₆H₃Cl₃O
  • Description:
    3,4,5-Trichlorophenol

Specifications

About this item