Potassium chloride saturated with silver chloride aqueous solution 4 mol/l (4 N) in aqueous solution
REGEEFS4AC
: REAGECON
:
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
:
:74.55 g/mol
:1.13 g/cm³ (20 °C)
:Ambient
:231-211-8
:7447-40-7
:KCl+AgCl
:MFCD00011360