ContactUsLinkComponent
Sodium azide tablets for milk preservation, Sigma-Aldrich®
Sodium azide tablets for milk preservation, Sigma-Aldrich®
  1.06687.0001
 :  Merck
 :  
Sodium azide tablets for milk preservation, Sigma-Aldrich®
  1.06687.0001
 :  Merck
 :  
  • Pk:
    5.000 Tab.
  • Description:
    Sodium azide tablets
Simple to use tablet for preservation of milk samples prior to testing. Each 100 mg tablet contains approximately 8 mg of sodium azide.
 : Packed in plastic can.