Order Entry
Switzerland
ContactUsLinkComponent
Scraper, SP Bel-Art
Scraper, SP Bel-Art
Catalog # BELAF368200001
undefined
Scraper, SP Bel-Art
Catalog # BELAF368200001

Specifications

  • Description:
    Triangular scraper, width 111 mm, with tapered blade, white
  • Length:
    248 mm
  • Pk:
    1

Specifications

About this item

PP, white.

  • Rigid, chisel-edged, triangular blade is tapered and fans out to a width of 11,1 cm (4⅜")
  • Handle is shaped for a comfortable and secure grip
  • Steam autoclavable at 121 °C
  • Complies with FDA requirements for direct contact with foods, drugs and cosmetics