ContactUsLinkComponent
Cub - dry ice maker, Frigimat®, SP Bel-Art
Cub - dry ice maker, Frigimat®, SP Bel-Art
Catalog # BELAF388740000
undefined
Cub - dry ice maker, Frigimat®, SP Bel-Art
Catalog # BELAF388740000
Specifications
  • Weight:
    2,2 kg
  • Description:
    Cub - dry ice maker
  • W×D×H:
    143×192×152 mm
  • Pk:
    1
Specifications
About this item
This compact device can be attached to a liquid CO₂ cylinder (not included) equipped with a dip tube to produce a ready-to-use 250 to 350 g block of dry ice in only a few minutes.
  • Lightweight body allows safe transport of the unit and contents using appropriate gloves
  • Operates at very low pressure
  • Rugged nickel-plated 'pigtail' connects it to the CO₂ cylinder valve which is used to regulate gas flow
Dry ice production can be viewed through the transparent polycarbonate body that secures without clamps, screws or tools ensuring easy removal of the solid block of dry ice and disconnection from the CO₂ cylinder.
Delivery: It includes nickel-plated copper connecting tube (1,8 m) with standard CGA 320 thread coupling for easy attachment to a CO₂ cylinder.