ContactUsLinkComponent
Bosshead, SP Bel-Art
Bosshead, SP Bel-Art
Catalog # BELAH183500000
undefined
Bosshead, SP Bel-Art
Catalog # BELAH183500000
Specifications
  • Material:
    Stainless steel
  • Pk:
    1
  • For rod Ø:
    13 mm
Specifications
About this item
Stainless steel. This open-sided bosshead accepts rods up to 13 mm diameter.
Holds rods at right angles to one another with flat wing-head clamping screws.