Pipette stand, Poxygrid®, SP Bel-Art
Catalog # BELAH189620006
Supplier: Bel-Art Products, a Part of SP
Specifications
- For:5 pipettes
- Description:Pipette stand with notch shaped wells
- Material:Stainless steel
- Seq:01
- W×D×H:229×146×197 mm
- Pk:1
Specifications
About this item
Pipette stand made of stainless steel.
- Uncoated stainless steel is corrosion resistant and easy to clean
- Supports 5 pipettors
- Suitable for most popular brands including Gilson Pipetman® pipettor