Order Entry
Switzerland
ContactUsLinkComponent
Rack for cuvettes, No-Wire™, SP Bel-Art
Rack for cuvettes, No-Wire™, SP Bel-Art
Catalog # 634-8290
undefined
Rack for cuvettes, No-Wire™, SP Bel-Art
Catalog # 634-8290

Specifications

  • For:
    42 (6×7)
  • W×D×H:
    129×104×43 mm
  • Pk:
    1

Specifications

About this item

These PP rack provides gentle no-scratch support for fragile and expensive spectrophotometer cuvettes.

  • Holds forty-two 10 mm light path cuvettes
  • 6×7 row layout
  • Steam autoclavable at 121 °C