ContactUsLinkComponent
Screw thread vials, Target®
  NSCAC4010-V1
undefined
Screw thread vials, Target®
  NSCAC4010-V1
  • Cap included:
    No
  • Ø×H:
    12×32 mm
  • Capacity:
    1,50 ml
  • Colour:
    Clear
  • Description:
    Vials, tapered base, with 150 µl reservoir
  • Material:
    Glass
  • Seq:
    11
  • Pk:
    100
Vials feature a 40% larger opening than standard vials and a uniformly flat bottom for security with inserts. Made from Type I, Class A borosilicate glass. Target I-D™ vials have a ceramic write-on ID patch with graduations at 0,5, 1,0 and 1,5 ml. Silanised vials help to limit protein adsorption.
Inserts are designed for wide opening 2 ml vials. Flat bottom MicroSert™ inserts are made from borosilicate glass. Conical-style Polyspring® inserts are available in borosilicate glass or PP.
Screw caps and septa