Order Entry
Switzerland
ContactUsLinkComponent
Selenium standard solution, concentrate, 1000 mg Se in 5% nitric acid (from SeO₂) Titrisol® standard for AAS, Supelco®
Selenium standard solution, concentrate, 1000 mg Se in 5% nitric acid (from SeO₂) Titrisol® standard for AAS, Supelco®
Katalognr. 1.09915.0001
Supplier:  Merck
CAS Number:  
Selenium standard solution, concentrate, 1000 mg Se in 5% nitric acid (from SeO₂) Titrisol® standard for AAS, Supelco®
Katalognr. 1.09915.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Element:
    Selenium
  • Symbol:
    Se

Specifications

About this item

Certifications: Traceable to SRM from NIST