Order Entry
Switzerland
ContactUsLinkComponent
Pesticide reference standard, Triazophos (O,O-Diethyl-O-1-phenyl-1H-1,2,4-triazol-3-yl phosphorothioate) 100 µg/ml in acetone, Dr Ehrenstorfer
Pesticide reference standard, Triazophos (O,O-Diethyl-O-1-phenyl-1H-1,2,4-triazol-3-yl phosphorothioate) 100 µg/ml in acetone, Dr Ehrenstorfer
Catalog # EHERXA17650000AC
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Triazophos (O,O-Diethyl-O-1-phenyl-1H-1,2,4-triazol-3-yl phosphorothioate) 100 µg/ml in acetone, Dr Ehrenstorfer
Catalog # EHERXA17650000AC
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    24017-47-8
  • Description:
    Triazophos (O,O-Diethyl-O-1-phenyl-1H-1,2,4-triazol-3-yl phosphorothioate) 100 µg/ml in acetone
  • Formula:
    C₁₂H₁₆N₃O₃PS
  • Description:
    Triazophos (O,O-Diethyl-O-1-phenyl-1H-1,2,4-triazol-3-yl phosphorothioate)

Specifications

About this item