
Bis(4-hydroxyphenyl)sulphide for synthesis, Sigma-Aldrich®
Catalog # 8.14209.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:149 - 152 °C
- Molar Mass:218.26 g/mol
Specification Test Results
About this item
:
:218.28 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:220-197-9
UN:3261
:GHS07
Storage Temperature:Ambient
CAS Number:2664-63-3
Formula:C₁₂H₁₀O₂S
MDL Number:MFCD00002349