(4-Pyridylthio)acetic acid for synthesis
Catalog # 8.14029.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:255 - 258 °C decomposes
- Molar Mass:169.19 g/mol
- Bulk Density:340 - 390 kg/m3
- Ignition Point:305 °C
Specification Test Results
About this item
:
:169.2 g/mol
ADR:6.1,I
:M-Clarity™ quality level = MQ200
EINECS:233-765-6
UN:2811
:GHS07
Storage Temperature:Ambient
CAS Number:10351-19-6
Formula:C₇H₇NO₂S
MDL Number:MFCD00006424