Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Nonadecanoic acid methyl ester

Nonadecanoic acid methyl ester

Supplier:   MATREYA
Nonadecanoic acid methyl ester
Supplier:  MATREYA
Certificates
Nonadecanoic acid methyl ester

1 Options Available Below

Product Details & Documents
Synonyms:  Nonadecanoic acid methyll ester, Methyl Nonadecanoate
  • CAS Number:1731-94-8
  • Formula:C₂₀H₄₀O₂
  • MDL Number:MFCD00009011
  • Molecular Weight:312.54 g/mol
  • EINECS:217-056-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg

  • Glass bottle for solids
    100 mg
    Discontinued
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.