Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Pesticide reference standard, Dieldrin 100 µg/ml in n-nonane, CIL
Pesticide reference standard, Dieldrin 100 µg/ml in n-nonane, CIL
PROOCILULM72301.2
:
LGC Standards PROMOCHEM
:
Pesticide reference standard, Dieldrin 100 µg/ml in n-nonane, CIL
PROOCILULM72301.2
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
1,2 mL
CAS No.:
60-57-1
Description:
Dieldrin 100 µg/ml in n-nonane
Formula:
C₁₂H₈Cl₆O
Description:
Dieldrin