Order Entry
Switzerland
ContactUsLinkComponent
 
Sodium standard solution, 1000 mg/L Na in water standard for ion chromatography
  REGEICCS04
 :  REAGECON
 :  
Sodium standard solution, 1000 mg/L Na in water standard for ion chromatography
  REGEICCS04
 :  REAGECON
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Sodium
  • Symbol:
    Na