
Ammoniumhexachlororhodat(III) hydrat ≥99,995% (Metall-Basis), Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15336-18-2
:
Formula:H₁₂Cl₆N₃Rh·H₂O
MDL Number:MFCD00003394
:369.74 g/mol (anhy)
Melting Pt: Zersetzung
EINECS:239-364-2
:GHS05
Storage Temperature:Raumtemperatur
Related Information
Frequently Bought Together