Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

8-Hydroxy-2-tetralone

Glass bottle for solids, 1 g
Catalog # MOLE57291624-1G
Marketsource
Supplier:   Molekula
8-Hydroxy-2-tetralone
Catalog # MOLE57291624-1G
Supplier:  Molekula
Marketsource
CAS Number:  53568-05-1
8-Hydroxy-2-tetralone

Glass bottle for solids, 1 g

CAS Number:  53568-05-1
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  8-Hydroxy-3,4-dihydronaphthalen-2(1H)-one
  • CAS Number:53568-05-1
  • Formula:C₁₀H₁₀O₂
  • MDL Number:MFCD01314169
  • Molecular Weight:162.19 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications