Herbicide reference standard, Desethyl atrazine 100 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERXA10331000CY
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:6190-65-4
- Description:Desethyl atrazine 100 µg/ml in cyclohexane
- Formula:C₆H₁₀ClN₅
- Description:Desethyl atrazine