



Tributylborane 1 M in THF, AcroSeal®
:122-56-5
:
:C₁₂H₂₇B
:MFCD00009423
:182.16 g/mol
:0.847 g/cm³
:–20 °C
:Ambient
:204-554-6
:3392
:4.2,
:GHS05
:GHS02
:GHS07
:GHS08
- Appearance:Clear colorless to light yellow Solution
- Strength:0.95 to 1.10 M
- Loss on drying:≤0.5 % (105°C, 4 hr)
- Residue after ignition:≤0.1 %
- Titration with HClO4:99.0 to 101.0 %
Frequently Bought Together