Supelco® PESTANAL® Organic Reference Standard, Simazine Solution
Simazine solution, Analytical standard, Glass bottle, 2 mL
Catalog # 36588-2ML
Supplier: Merck
Supelco® PESTANAL® Organic Reference Standard, Simazine Solution
Catalog # 36588-2ML
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2 mL
- Standards:Analytical standard
- Description:Simazine solution
- Cat. No.:36588-2ML
- M-Clarity quality level:MQ100
- Formula:C₇H₁₂ClN₅
- Storage conditions:2...8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- MDL No.:MFCD00023174
- Flash point:11 °C
- Molecular weight:201,66
- CAS No.:122-34-9
- Shipping conditions:STD



