Herbicide reference standard, Simazine (2-Chloro-4,6-bis(ethylamino)-1,3,5-triazine) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERXA16950000AL
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:122-34-9
- Description:Simazine (2-Chloro-4,6-bis(ethylamino)-1,3,5-triazine) 100 µg/ml in acetonitrile
- Formula:C₇H₁₂ClN₅
- Description:Simazine (2-Chloro-4,6-bis(ethylamino)-1,3,5-triazine)