Order Entry
Switzerland
ContactUsLinkComponent
Herbicide reference standard, Trifluralin 100 µg/ml in n-nonane, CIL
Herbicide reference standard, Trifluralin 100 µg/ml in n-nonane, CIL
Catalog # PROOCILULM72361.2
CAS Number:  
Herbicide reference standard, Trifluralin 100 µg/ml in n-nonane, CIL
Catalog # PROOCILULM72361.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    1582-09-8
  • Description:
    Trifluralin 100 µg/ml in n-nonane
  • Formula:
    C₁₃H₁₆F₃N₃O₄
  • Description:
    Trifluralin

Specifications

About this item