Herbicide reference standard, Dinoterb (2-tert-Butyl-4,6-dinitrophenol) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA12830000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:1420-07-1
- Description:Dinoterb (2-tert-Butyl-4,6-dinitrophenol) 100 µg/ml in acetonitrile
- Formula:C₁₀H₁₂N₂O₅
- Description:Dinoterb (2-tert-Butyl-4,6-dinitrophenol)