Order Entry
Switzerland
ContactUsLinkComponent
 
Tantalum standard solution, 1000 mg/L Ta in water (from (NH₄)₂TaF₇) Certipur® standard for ICP, Supelco®
Catalog # 1.70356.0100
Supplier:  Merck
CAS Number:  
undefined
Tantalum standard solution, 1000 mg/L Ta in water (from (NH₄)₂TaF₇) Certipur® standard for ICP, Supelco®
Catalog # 1.70356.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Tantalum
  • Symbol:
    Ta

Specifications

About this item

Certifications: Traceable to SRM from NIST