Order Entry
Switzerland
ContactUsLinkComponent
 
Rhenium standard solution, for AAS, 1 mg/ml Re in water
Catalog # ACRO196221000
CAS Number:  
Rhenium standard solution, for AAS, 1 mg/ml Re in water
Catalog # ACRO196221000
CAS Number:  

Specifications

  • Concentration:
    1 mg/ml
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Environmentally Preferable:
  • Element:
    Rhenium
  • Symbol:
    Re

Specifications

About this item