Europium standard solution, 1000 mg/L Eu in 5% nitric acid (from Eu?O?), standard for ICP, Specpure®
Molecular Formula: Eu₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010992
: Traceable to NIST
- Environmentally Preferable:
Frequently Bought Together