Order Entry
Switzerland
ContactUsLinkComponent
Yttrium standard solution, 10 mg/L Y in 5% nitric acid (from Y₂O₃), standard for ICP, Specpure®
Yttrium standard solution, 10 mg/L Y in 5% nitric acid (from Y₂O₃), standard for ICP, Specpure®
Catalog # 45268.AE
CAS Number:  
Yttrium standard solution, 10 mg/L Y in 5% nitric acid (from Y₂O₃), standard for ICP, Specpure®
Catalog # 45268.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Environmentally Preferable:
  • Element:
    Yttrium
  • Symbol:
    Y

Specifications

About this item

Certifications: Traceable to NIST