Order Entry
Switzerland
ContactUsLinkComponent
Ruthenium standard solution, 1,000 mg/L Ru in 5% hydrochloric acid standard for AAS
Ruthenium standard solution, 1,000 mg/L Ru in 5% hydrochloric acid standard for AAS
Catalog # ACRO196251000
CAS Number:  
Ruthenium standard solution, 1,000 mg/L Ru in 5% hydrochloric acid standard for AAS
Catalog # ACRO196251000
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Ruthenium
  • Symbol:
    Ru

Specifications

About this item