Ruthenium standard solution, 10000 mg/L Ru in 20% hydrochloric acid (from RuCl₃), standard for ICP, Specpure®
Nome del fornitore: Thermo Fisher Scientific
Certificati
Informazioni su questo articolo
Molecular Formula: RuCl₃ in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011207
Certifications: Traceable to NIST
Specifiche
- MDL No.:MFCD00011207
Specifiche
Frequently Bought Together