Order Entry
Switzerland
ContactUsLinkComponent
Pesticide reference standard, Pentachlorophenol 100 µg/ml in n-nonane, CIL
Pesticide reference standard, Pentachlorophenol 100 µg/ml in n-nonane, CIL
  PROOCILULM68941.2
 :  
Pesticide reference standard, Pentachlorophenol 100 µg/ml in n-nonane, CIL
  PROOCILULM68941.2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    87-86-5
  • Description:
    Pentachlorophenol 100 µg/ml in n-nonane
  • Formula:
    C₆HCl₅O
  • Description:
    Pentachlorophenol