Magnesium standard solution, 1000 mg/L Mg in 5% nitric acid (from Mg), standard for ICP, Specpure®
Molecular Formula: Mg in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085308
: Traceable to NIST
- MDL No.:MFCD00085308
Frequently Bought Together