Magnesium standard solution, 10000 mg/L Mg in 5% nitric acid (from Mg), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Mg In 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085308
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00085308
Specifications
Related Information
Frequently Bought Together