Order Entry
Switzerland
ContactUsLinkComponent
 
Flammable
Irritant
Health
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Catalog # 9827-02
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
Health
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Catalog # 9827-02
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 0.01
  • UV Absorbance - 260 - 400 nm:
    ≤0.01
  • UV Absorbance - 230 nm:
    ≤0.15
  • UV Absorbance - 220 nm:
    ≤0.30
  • Assay (CH₃CHOH CH₃):
    ≥99.8 %
  • Residue after Evaporation:
    ≤1.0 ppm
  • Water (H₂O):
    ≤0.06 %
  • LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine):
    ≤50 ppb
  • Trace Impurities - Aluminum (Al):
    ≤50.0 ppb
  • Trace Impurities - * Calcium (Ca):
    ≤50.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤50.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤50.0 ppb
  • Trace Impurities - Potassium (K):
    ≤50 ppb
  • Trace Impurities - * Sodium (Na):
    ≤50.0 ppb

Specification Test Results

About this item

 :
 :60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
 :GHS02
 :GHS07
 :GHS08
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674