Order Entry
Switzerland
ContactUsLinkComponent
 
Phosphorus standard solution, 1,000 mg/L P in water standard for AAS
  ACRO196185000
 :  
Phosphorus standard solution, 1,000 mg/L P in water standard for AAS
  ACRO196185000
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Phosphorus
  • Symbol:
    P