Order Entry
Switzerland
ContactUsLinkComponent
Yttrium standard solution, 10000 mg/L Y in dil. nitric acid standard for ICP
Yttrium standard solution, 10000 mg/L Y in dil. nitric acid standard for ICP
Catalog # REGEPY2B2
Supplier:  REAGECON
CAS Number:  
Yttrium standard solution, 10000 mg/L Y in dil. nitric acid standard for ICP
Catalog # REGEPY2B2
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Element:
    Yttrium
  • Symbol:
    Y

Specifications

About this item