Yttrium standard solution, 10,000 µg/l Y in 5% nitric acid (from Y₂O₃) standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Y₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011468
Certifications: Traceable to NIST
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together