Wolfram-Standardlösung, 100000 mg/L W in Wasser (aus (NH4)2WO4) Specpure® Standard für die ICP
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: (NH₄)₂WO₄ in H₂O
Physical Form: Liquid
MDL No.: MFCD00011461
Certifications: Rückführbar auf NIST
Specifications
- MDL No.:MFCD00011461
Specifications
Related Information
Frequently Bought Together