Order Entry
Switzerland
ContactUsLinkComponent
Vanadium standard solution, 1000 mg/L V in 0.5 M nitric acid standard for AAS
Vanadium standard solution, 1000 mg/L V in 0.5 M nitric acid standard for AAS
Catalog # REGEAAV-H
Supplier:  REAGECON
CAS Number:  
Vanadium standard solution, 1000 mg/L V in 0.5 M nitric acid standard for AAS
Catalog # REGEAAV-H
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Vanadium
  • Symbol:
    V

Specifications

About this item