Order Entry
Switzerland
ContactUsLinkComponent
Ammonium hydrogen carbonate 21,30 - 21,73% (NH₃ basis), powder, BAKER ANALYZED®, J.T.Baker®
Ammonium hydrogen carbonate 21,30 - 21,73% (NH₃ basis), powder, BAKER ANALYZED®, J.T.Baker®
Catalog # 3003-01
Supplier:  Avantor
CAS Number:  
undefined
Ammonium hydrogen carbonate 21,30 - 21,73% (NH₃ basis), powder, BAKER ANALYZED®, J.T.Baker®
Catalog # 3003-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    21.30 - 21.73 %
  • Assay (as NH₃):
    21.30 - 21.73 %
  • pH of 5% Solution at 25°C:
    7.0 - 8.0
  • Nonvolatile Matter:
    ≤0.02 %
  • Insoluble Matter:
    ≤0.005 %
  • Nitrate (NO₃):
    ≤0.002 %
  • Sulfur Compounds (as SO₄):
    ≤0.002 %
  • Chloride (Cl):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

About this item

Synonyms: Ammonium bicarbonate
 :
 :79.06 g/mol
Merck Index:13,00497
Melting Pt:35 °C
Density:1.57 g/cm³ (20 °C)
EINECS:213-911-5
 :GHS07
CAS Number:1066-33-7
Formula:NH₄HCO₃
MDL Number:MFCD00012138