Order Entry
Switzerland
ContactUsLinkComponent
Sodium standard solution, 1000 mg/L Na in 0,5 M nitric acid standard for AAS
Sodium standard solution, 1000 mg/L Na in 0,5 M nitric acid standard for AAS
Catalog # REGEAANAH
Supplier:  REAGECON
CAS Number:  
Sodium standard solution, 1000 mg/L Na in 0,5 M nitric acid standard for AAS
Catalog # REGEAANAH
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Sodium
  • Symbol:
    Na

Specifications

About this item