Order Entry
Switzerland
ContactUsLinkComponent
 
Silver standard solution, 1000 mg/L Ag in 0,5 M nitric acid standard for AAS
  REGEAAAGH
 :  REAGECON
 :  
Silver standard solution, 1000 mg/L Ag in 0,5 M nitric acid standard for AAS
  REGEAAAGH
 :  REAGECON
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Silver
  • Symbol:
    Ag