Order Entry
Switzerland
ContactUsLinkComponent
Magnesium standard solution, 10000 mg/L Mg in dil. nitric acid (from Mg(NO₃)₂) Certipur® standard for ICP, Supelco®
Magnesium standard solution, 10000 mg/L Mg in dil. nitric acid (from Mg(NO₃)₂) Certipur® standard for ICP, Supelco®
Catalog # 1.70379.0100
Supplier:  Merck
CAS Number:  
Magnesium standard solution, 10000 mg/L Mg in dil. nitric acid (from Mg(NO₃)₂) Certipur® standard for ICP, Supelco®
Catalog # 1.70379.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Magnesium
  • Symbol:
    Mg

Specifications

About this item

Certifications: Traceable to SRM from NIST