Mortars without pestles, SP Bel-Art
Anbieter: Bel-Art Products, a Part of SP
Zertifikate
Über diesen Artikel
Liquid nitrogen cooled mortars without pestles, for grinding frozen samples.
- Insulated handle is removable to save space; 10,2cm long
- Three suction cup feet stabilise the unit on the workbench
- Recommended size pestle is any size appropriate for a 275ml mortar (not included)
Grind frozen samples without liquid nitrogen hazards. This 304 stainless steel mortar is suspended by three brackets above a stainless steel bowl which holds liquid nitrogen. The bowl is nested in a heavily insulated blue polyethylene housing.
Frequently Bought Together