Eco stopcock plugs, with taper 1:5
Supplier: Bohlender
Certificates
About this item
Made of PFTE, white, with grip and nut of PP, red.
- Universal chemical resistance
- Temperature resistance: from –10 to +120 °C
- Taper: 1:5
- With or without bore
- Washer included
Frequently Bought Together